Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024769.3 | NP_079045.1 | 6, 7 | 1181 | ||
GenBank mRNA | AK026068.1 | - | 6, 7 | 1157 | ||
AK304843.1 | - | 6, 7 | 901 | |||
AY326422.1 | - | 6, 7 | 1005 | |||
AY358340.1 | - | 6, 7 | 1005 | |||
BC009371.2 | - | 6, 7 | 827 | 4122648 |