Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM15B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013286.4 | NP_037418.3 | 1 | 3013 | ||
GenBank mRNA | AL831838.1 | - | 1 | 2419 | ||
BC001367.2 | - | 1 | 1555 | |||
BC121184.2 | - | 2 | 894 | |||
BC139836.1 | - | 2 | 2224 | 40123447 | ||
L13434.1 | - | 1 | 2770 |