Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRAMD1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172105.1 | NP_001165576.1 | 7 | 1045 | ||
NM_017577.4 | NP_060047.3 | 12 | 1318 | |||
XM_005247546.2 | XP_005247603.1 | 11 | 1234 | |||
XM_005247547.1 | XP_005247604.1 | 12 | 1280 | |||
XM_011512930.1 | XP_011511232.1 | 12 | 1313 | |||
XM_011512931.1 | XP_011511233.1 | 7 | 1102 | |||
XM_017006646.1 | XP_016862135.1 | 11 | 1232 | |||
XM_017006647.1 | XP_016862136.1 | 13 | 1396 | |||
XM_017006648.1 | XP_016862137.1 | 8 | 1125 | |||
XM_017006649.1 | XP_016862138.1 | 6 | 876 | |||
GenBank mRNA | AK093181.1 | - | 7 | 837 | ||
AK097783.1 | - | 13 | 1402 | |||
AK292846.1 | - | 12 | 1254 | |||
AK292964.1 | - | 7 | 960 | |||
AK293884.1 | - | 10 | 1030 | |||
AK304142.1 | - | 7 | 1045 | |||
AL133661.1 | - | 3 | 1534 | |||
AY358725.1 | - | 7 | 998 | |||
BC028972.2 | - | 4 | 322 | |||
BC035040.1 | - | 12 | 1324 | 4829130 |