Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PBRM1 SIRNA Assays ›
Gene Symbol:
PBRM1
Gene Name:
polybromo 1
Gene Aliases:
BAF180, PB1
Chromosome Location:
Chr.3: 52545352 - 52685933 on Build GRCh38
UniGene ID:
Hs.189920
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
125128
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PBRM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_018313.4
NP_060783.3
2
229
XM_005265279.4
XP_005265336.1
3
457
XM_005265280.3
XP_005265337.1
2
274
XM_005265282.3
XP_005265339.1
3
416
XM_005265283.3
XP_005265340.1
2
271
XM_011533900.2
XP_011532202.1
5
623
XM_011533902.2
XP_011532204.1
4
555
XM_011533903.2
XP_011532205.1
4
560
XM_017006725.1
XP_016862214.1
4
503
XM_017006726.1
XP_016862215.1
5
613
XM_017006727.1
XP_016862216.1
5
558
XM_017006728.1
XP_016862217.1
4
554
XM_017006729.1
XP_016862218.1
4
962
XM_017006730.1
XP_016862219.1
5
549
XM_017006731.1
XP_016862220.1
4
532
XM_017006732.1
XP_016862221.1
4
555
XM_017006733.1
XP_016862222.1
4
949
XM_017006734.1
XP_016862223.1
4
522
XM_017006735.1
XP_016862224.1
4
961
XM_017006736.1
XP_016862225.1
4
962
XM_017006737.1
XP_016862226.1
3
836
XM_017006738.1
XP_016862227.1
3
457
XM_017006739.1
XP_016862228.1
3
833
XM_017006740.1
XP_016862229.1
3
374
XM_017006741.1
XP_016862230.1
4
537
XM_017006742.1
XP_016862231.1
4
560
XM_017006743.1
XP_016862232.1
4
962
XM_017006744.1
XP_016862233.1
4
526
XM_017006745.1
XP_016862234.1
3
835
XM_017006746.1
XP_016862235.1
3
832
XM_017006747.1
XP_016862236.1
4
963
XM_017006748.1
XP_016862237.1
3
835
XM_017006749.1
XP_016862238.1
3
401
XM_017006750.1
XP_016862239.1
2
289
XM_017006751.1
XP_016862240.1
4
537
XM_017006752.1
XP_016862241.1
4
962
XM_017006753.1
XP_016862242.1
4
961
XM_017006754.1
XP_016862243.1
4
555
XM_017006755.1
XP_016862244.1
4
537
XM_017006756.1
XP_016862245.1
4
537
XM_017006757.1
XP_016862246.1
2
260
XM_017006758.1
XP_016862247.1
2
391
XM_017006759.1
XP_016862248.1
4
537
XM_017006760.1
XP_016862249.1
4
555
XM_017006761.1
XP_016862250.1
4
564
XM_017006762.1
XP_016862251.1
4
555
XM_017006763.1
XP_016862252.1
5
614
XM_017006764.1
XP_016862253.1
4
537
XM_017006765.1
XP_016862254.1
3
416
XM_017006766.1
XP_016862255.1
4
544
GenBank mRNA
AA047886.1
-
1
131
AF177387.1
-
1
118
AF197569.1
-
2
211
AF225870.1
-
1
118
AF225871.1
-
1
118
AF225872.1
-
1
118
AY281068.1
-
1
117
BC115009.1
-
1
133
7962003
BC115010.1
-
1
134
BC115011.1
-
1
134
7962010
BC129934.1
-
1
118
40082629
BC129935.1
-
1
118
40082630
BE672944.1
-
2
229
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
mitotic nuclear division
negative regulation of cell proliferation
covalent chromatin modification
DNA binding
chromatin binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-f2ldd:80/10.239.165.14:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0