Gene Symbol: C3orf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291941.1 | NP_001278870.1 | 6 | 590 | ||
NM_001291942.1 | NP_001278871.1 | 5 | 478 | |||
NM_020685.4 | NP_065736.1 | 6 | 697 | |||
XM_017006931.1 | XP_016862420.1 | 6 | 1073 | |||
GenBank mRNA | AF236158.1 | - | 6 | 527 | ||
BC017772.2 | - | 5 | 507 | 4307022 | ||
BU568674.1 | - | 432 |