Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002974.3 | NP_002965.1 | 8 | 1362 | ||
NM_175041.1 | NP_778206.1 | 8 | 1299 | |||
XM_011526138.1 | XP_011524440.1 | 8 | 1372 | |||
GenBank mRNA | AK292212.1 | - | 8 | 1362 | ||
BC017401.1 | - | 8 | 1302 | 4047303 |