Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIS3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152383.4 | NP_689596.4 | 14 | 1989 | ||
NR_046476.1 | 14 | 1989 | ||||
NR_046477.1 | 13 | 1965 | ||||
GenBank mRNA | AK295363.1 | - | 14 | 1859 | ||
AL834174.1 | - | 15 | 2683 | |||
BC026166.1 | - | 14 | 1893 | 4822064 | ||
BC036113.1 | - | 14 | 1888 | 5297794 | ||
BG746662.1 | - | 2 | 196 | |||
BX648325.1 | - | 14 | 1856 |