Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IZUMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321864.1 | NP_001308793.1 | 5 | 417 | ||
NM_001321865.1 | NP_001308794.1 | 5 | 543 | |||
NM_182575.2 | NP_872381.2 | 6 | 1018 | |||
NR_135832.1 | 5 | 476 | ||||
XM_005258793.4 | XP_005258850.1 | 6 | 751 | |||
XM_011526812.2 | XP_011525114.1 | 6 | 1098 | |||
XM_011526813.2 | XP_011525115.1 | 6 | 947 | |||
XM_011526815.2 | XP_011525117.1 | 5 | 1661 | |||
XM_017026646.1 | XP_016882135.1 | 5 | 1522 | |||
GenBank mRNA | AB195682.1 | - | 6 | 876 | ||
AK301750.1 | - | 5 | 1661 | |||
AY552609.1 | - | 5 | 470 | |||
AY552610.1 | - | 5 | 403 | |||
BC034769.1 | - | 6 | 1018 | 5167853 |