Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LONRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198461.3 | NP_940863.3 | 9 | 2370 | ||
GenBank mRNA | AK127206.1 | - | 9 | 1072 | ||
BC101662.1 | - | 8 | 1029 | 8069168 | ||
BC101664.1 | - | 8 | 1029 | 8069170 | ||
BC143468.1 | - | 8 | 1029 | |||
DQ020495.1 | - | 9 | 2370 |