Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY358542.1 | - | 5, 6 | 679 | ||
DQ479397.1 | - | 5, 6 | 511 | |||
FJ584316.1 | - | 5, 6 | 505 | |||
FJ584320.1 | - | 5, 6 | 296 |