Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARFGEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006421.4 | NP_006412.2 | 19 | 3197 | ||
XM_005251134.4 | XP_005251191.1 | 20 | 3176 | |||
XM_005251135.3 | XP_005251192.1 | 19 | 3447 | |||
XM_005251136.3 | XP_005251193.1 | 19 | 3447 | |||
XM_011517442.2 | XP_011515744.1 | 20 | 3483 | |||
GenBank mRNA | AB209324.1 | - | 9 | 1242 | ||
AF084520.1 | - | 19 | 2807 | |||
AF111162.1 | - | 19 | 2948 |