Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM49B SIRNA Assays ›
Gene Symbol:
FAM49B
Gene Name:
family with sequence similarity 49 member B
Gene Aliases:
BM-009, L1
Chromosome Location:
Chr.8: 129839593 - 130016697 on Build GRCh38
UniGene ID:
Hs.126941
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
126945
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM49B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256763.1
NP_001243692.1
13
1611
NM_016623.4
NP_057707.3
12
1524
NR_046359.1
15
1877
NR_046360.1
15
1787
NR_046361.1
14
1777
XM_005250966.3
XP_005251023.1
11
1380
XM_011517108.2
XP_011515410.1
14
1673
XM_011517109.2
XP_011515411.1
14
1445
XM_011517110.2
XP_011515412.1
13
1585
XM_011517111.2
XP_011515413.1
13
1403
XM_011517112.2
XP_011515414.1
15
1628
XM_011517113.2
XP_011515415.1
14
1639
XM_011517114.2
XP_011515416.1
14
1545
XM_011517115.2
XP_011515417.1
13
1552
XM_011517116.2
XP_011515418.1
14
1586
XM_011517117.2
XP_011515419.1
13
1555
XM_011517118.2
XP_011515420.1
13
1499
XM_011517120.2
XP_011515422.1
12
1498
XM_011517121.2
XP_011515423.1
15
1510
XM_011517122.2
XP_011515424.1
13
1589
XM_011517123.2
XP_011515425.1
13
1362
XM_011517124.2
XP_011515426.1
12
1477
XM_011517125.2
XP_011515427.1
12
1416
XM_017013539.1
XP_016869028.1
15
1632
XM_017013540.1
XP_016869029.1
15
1491
XM_017013541.1
XP_016869030.1
13
1619
XM_017013542.1
XP_016869031.1
15
1669
XM_017013543.1
XP_016869032.1
15
1769
XM_017013544.1
XP_016869033.1
15
1551
XM_017013545.1
XP_016869034.1
16
1815
XM_017013546.1
XP_016869035.1
14
1582
XM_017013547.1
XP_016869036.1
16
1674
XM_017013548.1
XP_016869037.1
14
1678
XM_017013549.1
XP_016869038.1
14
1541
XM_017013550.1
XP_016869039.1
13
1592
XM_017013551.1
XP_016869040.1
15
1587
XM_017013552.1
XP_016869041.1
15
1632
XM_017013553.1
XP_016869042.1
14
1445
XM_017013554.1
XP_016869043.1
16
1657
XM_017013555.1
XP_016869044.1
13
1612
XM_017013556.1
XP_016869045.1
13
1358
XM_017013557.1
XP_016869046.1
14
1404
XM_017013558.1
XP_016869047.1
14
1549
XM_017013559.1
XP_016869048.1
14
1408
XM_017013560.1
XP_016869049.1
12
1523
XM_017013561.1
XP_016869050.1
12
1275
XM_017013562.1
XP_016869051.1
13
1462
GenBank mRNA
AF208851.1
-
14
1466
AK001226.1
-
8
1030
AK130372.1
-
12
1411
BC003599.1
-
13
1390
3501828
BC016345.1
-
14
1531
4041187
BC017297.2
-
15
1759
5017493
CR749628.1
-
11
4068
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-zp8kv:80/10.239.163.191:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0