Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF280C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017666.4 | NP_060136.1 | 19 | 3139 | ||
XM_006724765.3 | XP_006724828.1 | 18 | 2943 | |||
GenBank mRNA | AL834333.1 | - | 19 | 3070 | ||
BC051728.1 | - | 19 | 3090 | 4828435 |