Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018268.3 | NP_060738.2 | 13 | 1882 | ||
XM_005248551.4 | XP_005248608.1 | 11 | 1675 | |||
XM_011543505.2 | XP_011541807.1 | 13 | 1644 | |||
XM_017009606.1 | XP_016865095.1 | 13 | 2292 | |||
GenBank mRNA | AF115511.1 | - | 13 | 1629 | ||
AK001766.1 | - | 13 | 1828 | |||
AK021855.1 | - | 13 | 1668 | |||
AK023977.1 | - | 13 | 1584 | |||
AK293316.1 | - | 11 | 1367 | |||
AK300058.1 | - | 11 | 1493 | |||
AL834138.1 | - | 7 | 2563 | |||
BC040241.1 | - | 13 | 1634 | 3447269 |