Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175061.3 | NP_778231.2 | 5 | 828 | ||
XM_006715656.1 | XP_006715719.1 | 6 | 704 | |||
XM_017011831.1 | XP_016867320.1 | 5 | 596 | |||
XM_017011832.1 | XP_016867321.1 | 5 | 528 | |||
GenBank mRNA | AK000711.1 | - | 2 | 55 | ||
AK091311.1 | - | 5 | 753 | |||
AL832491.1 | - | 3 | 406 | |||
AL834234.1 | - | 4 | 547 | |||
AY372319.1 | - | 5 | 606 | |||
BC042441.1 | - | 6 | 616 | 4831063 | ||
BC047229.1 | - | 5 | 615 |