Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080870.3 | NP_543146.2 | 2 | 3857 | ||
GenBank mRNA | AB064272.1 | - | 1 | 385 | ||
AL833738.1 | - | 1 | 1199 | |||
BC069477.1 | - | 1 | 383 | 7262259 | ||
BC101661.1 | - | 1 | 448 | 8069167 | ||
BC101663.1 | - | 1 | 448 | 8069169 |