Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FREM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177704.1 | NP_001171175.1 | 12 | 1889 | ||
NM_144966.5 | NP_659403.4 | 36 | 7008 | |||
XM_005251382.3 | XP_005251439.1 | 35 | 7256 | |||
XM_005251384.4 | XP_005251441.1 | 14 | 2302 | |||
XM_006716729.3 | XP_006716792.1 | 15 | 2547 | |||
XM_011517758.2 | XP_011516060.1 | 12 | 1938 | |||
XM_017014316.1 | XP_016869805.1 | 35 | 7283 | |||
XM_017014317.1 | XP_016869806.1 | 36 | 6826 | |||
XM_017014318.1 | XP_016869807.1 | 36 | 7581 | |||
XM_017014319.1 | XP_016869808.1 | 37 | 7485 | |||
XM_017014320.1 | XP_016869809.1 | 36 | 7412 | |||
XM_017014321.1 | XP_016869810.1 | 35 | 7075 | |||
XM_017014322.1 | XP_016869811.1 | 35 | 6869 | |||
XM_017014323.1 | XP_016869812.1 | 35 | 6542 | |||
XM_017014324.1 | XP_016869813.1 | 34 | 7133 | |||
XM_017014326.1 | XP_016869815.1 | 34 | 6099 | |||
XM_017014327.1 | XP_016869816.1 | 31 | 5468 | |||
GenBank mRNA | AB160987.1 | - | 34 | 6192 | ||
AK058190.1 | - | 9 | 1058 | |||
BC031064.1 | - | 12 | 1889 | |||
BX537574.1 | - | 29 | 5583 | |||
BX641104.1 | - | 6 | 6513 |