Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEND6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152731.2 | NP_689944.2 | 7 | 2257 | ||
XM_005248889.1 | XP_005248946.1 | 5 | 1425 | |||
XM_011514346.2 | XP_011512648.1 | 5 | 1915 | |||
XM_017010406.1 | XP_016865895.1 | 7 | 1808 | |||
XM_017010407.1 | XP_016865896.1 | 7 | 1791 | |||
XM_017010408.1 | XP_016865897.1 | 8 | 2338 | |||
GenBank mRNA | AK054724.1 | - | 8 | 1839 | ||
AK315926.1 | - | 4 | 1221 | |||
BC022988.1 | - | 6 | 1652 | 4932760 |