Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STEAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040665.1 | NP_001035755.1 | 4 | 683 | ||
NM_001040666.1 | NP_001035756.1 | 4 | 701 | |||
NM_001244944.1 | NP_001231873.1 | 4 | 1020 | |||
NM_001244945.1 | NP_001231874.1 | 3 | 907 | |||
NM_001244946.1 | NP_001231875.1 | 4 | 683 | |||
NM_152999.3 | NP_694544.2 | 3 | 907 | |||
XM_006715921.3 | XP_006715984.1 | 4 | 1017 | |||
XM_017011953.1 | XP_016867442.1 | 4 | 847 | |||
XM_017011954.1 | XP_016867443.1 | 3 | 734 | |||
XM_017011955.1 | XP_016867444.1 | 4 | 820 | |||
XM_017011956.1 | XP_016867445.1 | 4 | 1018 | |||
XM_017011957.1 | XP_016867446.1 | 4 | 820 | |||
XM_017011958.1 | XP_016867447.1 | 3 | 4682 | |||
GenBank mRNA | AF455138.1 | - | 3 | 907 | ||
AF526382.1 | - | 4 | 701 | |||
AK092666.1 | - | 3 | 849 | |||
AK315043.1 | - | 4 | 821 | |||
AY008443.1 | - | 4 | 701 | |||
AY008444.1 | - | 4 | 683 | |||
AY358267.1 | - | 4 | 701 | |||
DQ656061.2 | - | 3 | 1877 | |||
DQ656062.2 | - | 3 | 871 |