Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP13-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181599.2 | NP_853630.2 | 1 | 511 | ||
GenBank mRNA | AB096939.1 | - | 1 | 511 | ||
AJ457066.1 | - | 1 | 379 | |||
BC113536.1 | - | 1 | 608 | 8322588 | ||
BC113538.1 | - | 1 | 608 | 8322590 |