Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MMS19 SIRNA Assays ›
Gene Symbol:
MMS19
Gene Name:
MMS19 homolog, cytosolic iron-sulfur assembly component
Gene Aliases:
MET18, MMS19L, hMMS19
Chromosome Location:
Chr.10: 97458324 - 97498794 on Build GRCh38
UniGene ID:
Hs.500721
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
130330
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MMS19
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289403.1
NP_001276332.1
4
524
NM_001289404.1
NP_001276333.1
3
475
NM_001289405.1
NP_001276334.1
5
406
NM_022362.4
NP_071757.4
4
524
XM_005270035.2
XP_005270092.1
3
281
XM_005270041.1
XP_005270098.1
3
298
XM_006717944.3
XP_006718007.2
4
318
XM_006717945.2
XP_006718008.1
4
362
XM_011540062.1
XP_011538364.1
4
256
XM_011540063.2
XP_011538365.1
4
362
XM_017016515.1
XP_016872004.1
5
430
XM_017016516.1
XP_016872005.1
5
430
XM_017016517.1
XP_016872006.1
5
430
XM_017016518.1
XP_016872007.1
4
318
XM_017016519.1
XP_016872008.1
5
430
XM_017016520.1
XP_016872009.1
4
318
XM_017016521.1
XP_016872010.1
4
318
XM_017016522.1
XP_016872011.1
4
422
XM_017016523.1
XP_016872012.1
3
306
XM_017016524.1
XP_016872013.1
3
281
XM_017016525.1
XP_016872014.1
3
187
XM_017016526.1
XP_016872015.1
3
262
XM_017016527.1
XP_016872016.1
3
281
XM_017016528.1
XP_016872017.1
4
362
XM_017016529.1
XP_016872018.1
5
430
XM_017016530.1
XP_016872019.1
4
422
XM_017016531.1
XP_016872020.1
3
281
XM_017016532.1
XP_016872021.1
3
187
XM_017016533.1
XP_016872022.1
2
138
XM_017016534.1
XP_016872023.1
4
362
XM_017016535.1
XP_016872024.1
4
362
GenBank mRNA
AF319947.1
-
5
372
AF357881.1
-
4
323
AJ306408.1
-
4
306
AK027710.1
-
3
158
AK225312.1
-
4
309
AK298995.1
-
4
362
AK304287.1
-
4
318
AK314374.1
-
4
322
BC080532.1
-
3
254
4889876
BC117129.1
-
4
322
40125680
DC382324.1
-
4
363
DC382327.1
-
4
362
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
phosphorelay signal transduction system
DNA metabolic process
DNA repair
nucleotide-excision repair
transcription, DNA-templated
cellular response to DNA damage stimulus
chromosome segregation
response to hormone
iron-sulfur cluster assembly
positive regulation of transcription, DNA-templated
transcription coactivator activity
protein binding
enzyme binding
receptor signaling complex scaffold activity
estrogen receptor binding
protein binding, bridging
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-68wbn:80/10.239.160.160:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0