Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160364.2 | NP_001153836.1 | 16 | 2703 | ||
NM_001324263.1 | NP_001311192.1 | 16 | 2592 | |||
NM_001324265.1 | NP_001311194.1 | 15 | 2449 | |||
NM_001324268.1 | NP_001311197.1 | 15 | 2457 | |||
NM_024847.4 | NP_079123.3 | 16 | 2605 | |||
NR_136733.1 | 15 | 2436 | ||||
GenBank mRNA | AK024893.1 | - | 12 | 1814 | ||
AY236498.1 | - | 16 | 2539 | |||
AY263165.1 | - | 16 | 2534 | |||
BC036205.1 | - | 16 | 2534 | 5261157 | ||
CR933669.1 | - | 16 | 2702 |