Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040451.2 | NP_001035541.1 | 8 | 1075 | ||
NM_001040452.2 | NP_001035542.1 | 9 | 886 | |||
NM_025158.4 | NP_079434.3 | 9 | 1061 | |||
XM_005265993.4 | XP_005266050.1 | 6 | 432 | |||
XM_006714921.3 | XP_006714984.2 | 8 | 1005 | |||
XM_006714922.3 | XP_006714985.1 | 6 | 865 | |||
XM_017009891.1 | XP_016865380.1 | 8 | 820 | |||
XM_017009892.1 | XP_016865381.1 | 7 | 858 | |||
XM_017009893.1 | XP_016865382.1 | 6 | 792 | |||
XM_017009894.1 | XP_016865383.1 | 10 | 1027 | |||
XM_017009895.1 | XP_016865384.1 | 9 | 1011 | |||
GenBank mRNA | AB209507.1 | - | 10 | 931 | ||
AF312367.1 | - | 9 | 1052 | |||
AF361055.2 | - | 9 | 1034 | |||
AK025904.1 | - | 3 | 124 | |||
AK075021.1 | - | 8 | 975 | |||
BC032571.1 | - | 9 | 805 | 5243223 |