Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC44A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178044.1 | NP_001171515.1 | 10 | 944 | ||
NM_001178045.1 | NP_001171516.1 | 11 | 1073 | |||
NM_025257.2 | NP_079533.2 | 11 | 1070 | |||
GenBank mRNA | AK027397.1 | - | 11 | 1021 | ||
AK222998.1 | - | 11 | 1021 | |||
AK223013.1 | - | 11 | 1021 | |||
AK300550.1 | - | 11 | 1073 | |||
AK301596.1 | - | 10 | 895 | |||
AK314673.1 | - | 11 | 1025 | |||
AL833009.1 | - | 7 | 763 | |||
BC014659.1 | - | 11 | 1009 | 4644272 | ||
DA924720.1 | - | 4 | 474 | |||
KU175229.1 | - | 11 | 1004 |