Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271287.1 | NP_001258216.1 | 4 | 474 | ||
NM_001271288.1 | NP_001258217.1 | 4 | 471 | |||
NM_001271318.1 | NP_001258247.1 | 6 | 634 | |||
NM_032434.3 | NP_115810.2 | 5 | 533 | |||
GenBank mRNA | AB058708.1 | - | 3 | 292 | ||
AK057028.1 | - | 5 | 443 | |||
AK294055.1 | - | 5 | 639 | |||
AK316011.1 | - | 6 | 494 | |||
AK316299.1 | - | 4 | 388 | |||
AL514772.3 | - | 5 | 450 | |||
BC043221.2 | - | 4 | 382 | 5295402 | ||
BC093041.1 | - | 4 | 401 | 6173398 | ||
BP251717.1 | - | 5 | 533 | |||
BX647996.1 | - | 4 | 380 |