siRNA Details
Gene Symbol: ZNF503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032772.5 | NP_116161.2 | 2 | 2898 | ||
GenBank mRNA | AK092897.1 | - | 1 | 2017 | ||
BC011625.2 | - | 2 | 2671 | 4053098 |

Gene Symbol: ZNF503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032772.5 | NP_116161.2 | 2 | 2898 | ||
GenBank mRNA | AK092897.1 | - | 1 | 2017 | ||
BC011625.2 | - | 2 | 2671 | 4053098 |