Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032804.5 | NP_116193.2 | 1 | 2155 | ||
GenBank mRNA | AK027453.1 | - | 1 | 1750 | ||
AK127694.1 | - | 1 | 2157 | |||
BC018660.1 | - | 1 | 2059 | 4104391 | ||
BC028589.2 | - | 1 | 2025 | 4838951 | ||
BC067740.1 | - | 1 | 1963 | 5295674 |