Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PJA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032396.2 | NP_001027568.1 | 2 | 933 | ||
NM_022368.4 | NP_071763.2 | 3 | 695 | |||
NM_145119.3 | NP_660095.1 | 2 | 1259 | |||
XM_005262292.1 | XP_005262349.1 | 2 | 921 | |||
XM_011531011.2 | XP_011529313.1 | 2 | 1066 | |||
GenBank mRNA | AF262024.1 | - | 1 | 1188 | ||
AF264620.1 | - | 3 | 610 | |||
AK021892.1 | - | 1 | 38 | |||
AK096611.1 | - | 2 | 1258 | |||
AK226171.1 | - | 2 | 840 | |||
AK290543.1 | - | 2 | 1159 | |||
AK293266.1 | - | 3 | 739 | |||
AK294632.1 | - | 3 | 772 | |||
BC039149.1 | - | 1 | 265 | |||
BC048323.1 | - | 1 | 105 | |||
BC058327.1 | - | 1 | 245 | |||
BC075803.1 | - | 2 | 1166 | 3913310 | ||
BC105051.1 | - | 1 | 948 | 8144054 | ||
BC105053.1 | - | 1 | 948 | 8144056 | ||
BC113035.1 | - | 1 | 269 |