Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAPBPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018009.4 | NP_060479.3 | 7 | 1660 | ||
XM_017019545.1 | XP_016875034.1 | 7 | 1808 | |||
XM_017019546.1 | XP_016875035.1 | 7 | 1488 | |||
GenBank mRNA | AK001005.1 | - | 7 | 1564 | ||
AK002056.2 | - | 7 | 1548 | |||
AK098035.1 | - | 8 | 1622 | |||
AK222956.1 | - | 7 | 1548 | |||
AK223163.1 | - | 7 | 1578 | |||
BC015017.2 | - | 7 | 1587 | 3916213 |