Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKAPK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003668.3 | NP_003659.2 | 7, 8 | 1093 | ||
NM_139078.2 | NP_620777.1 | 7, 8 | 1093 | |||
XM_017020125.1 | XP_016875614.1 | 7, 8 | 585 | |||
XM_017020126.1 | XP_016875615.1 | 5, 6 | 499 | |||
XM_017020127.1 | XP_016875616.1 | 6, 7 | 918 | |||
XM_017020128.1 | XP_016875617.1 | 6, 7 | 918 | |||
XM_017020129.1 | XP_016875618.1 | 6, 7 | 755 | |||
XM_017020130.1 | XP_016875619.1 | 6, 7 | 523 | |||
XM_017020131.1 | XP_016875620.1 | 6, 7 | 918 | |||
XM_017020132.1 | XP_016875621.1 | 7, 8 | 966 | |||
XM_017020133.1 | XP_016875622.1 | 6, 7 | 738 | |||
XM_017020134.1 | XP_016875623.1 | 6, 7 | 564 | |||
XM_017020135.1 | XP_016875624.1 | 5, 6 | 647 | |||
XM_017020136.1 | XP_016875625.1 | 6, 7 | 738 | |||
XM_017020137.1 | XP_016875626.1 | 5, 6 | 647 | |||
XM_017020138.1 | XP_016875627.1 | 1, 2 | 61 | |||
XM_017020139.1 | XP_016875628.1 | 5, 6 | 647 | |||
GenBank mRNA | AK122767.1 | - | 7, 8 | 1343 | ||
AL110301.1 | - | 2, 3 | 195 | |||
BC000833.1 | - | 6, 7 | 536 | |||
BC041049.1 | - | 7, 8 | 1080 | 4909021 | ||
BC047284.1 | - | 7, 8 | 761 | 4997201 |