Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXW5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018998.3 | NP_061871.1 | 5 | 753 | ||
XM_005266089.2 | XP_005266146.2 | 5 | 803 | |||
XM_011518789.1 | XP_011517091.1 | 5 | 803 | |||
XM_017014812.1 | XP_016870301.1 | 5 | 773 | |||
GenBank mRNA | AB209726.1 | - | 2 | 165 | ||
AK026081.1 | - | 5 | 617 | |||
AK315738.1 | - | 5 | 628 | |||
AL137631.1 | - | 1 | 16 | |||
BC004541.2 | - | 2 | 162 | |||
BC014130.1 | - | 5 | 649 | 4620371 | ||
BC014297.2 | - | 5 | 620 | 3958395 | ||
BC031100.1 | - | 4 | 768 | |||
BC035355.1 | - | 5 | 687 |