Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127244.1 | NP_001120716.1 | 4 | 3209 | ||
NM_001127245.1 | NP_001120717.1 | 3 | 2833 | |||
NM_019594.3 | NP_062540.2 | 4 | 2940 | |||
XM_005252095.2 | XP_005252152.1 | 4 | 3070 | |||
XM_005252096.3 | XP_005252153.1 | 4 | 4233 | |||
XM_006717186.2 | XP_006717249.1 | 5 | 3285 | |||
XM_011518863.1 | XP_011517165.1 | 4 | 2876 | |||
XM_011518864.2 | XP_011517166.1 | 5 | 2994 | |||
XM_011518865.2 | XP_011517167.1 | 3 | 2712 | |||
GenBank mRNA | AK001199.1 | - | 2 | 1078 | ||
AK027495.1 | - | 2 | 1876 | |||
AK074723.1 | - | 2 | 1912 | |||
AK289842.1 | - | 4 | 3209 | |||
AY358286.1 | - | 2 | 1973 | |||
BC000775.2 | - | 2 | 608 | |||
BC051322.1 | - | 4 | 2852 | 6250713 |