Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JPH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020655.3 | NP_065706.2 | 5 | 2646 | ||
GenBank mRNA | AK055486.1 | - | 4 | 1457 | ||
AK126663.1 | - | 4 | 2680 | |||
AK297215.1 | - | 2 | 616 | |||
AW954561.1 | - | 1 | 116 | |||
BC036533.2 | - | 5 | 2628 | 5264853 |