Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UNC79 SIRNA Assays ›
Gene Symbol:
UNC79
Gene Name:
unc-79 homolog (C. elegans)
Gene Aliases:
KIAA1409
Chromosome Location:
Chr.14: 93333219 - 93710473 on Build GRCh38
UniGene ID:
Hs.126561
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
133252
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UNC79
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_020818.3
NP_065869.3
45, 46
7333
XM_011537018.2
XP_011535320.1
47, 48
9966
XM_011537020.2
XP_011535322.1
46, 47
9900
XM_011537021.2
XP_011535323.1
47, 48
7517
XM_011537022.2
XP_011535324.1
47, 48
9810
XM_011537023.2
XP_011535325.1
47, 48
9807
XM_011537024.2
XP_011535326.1
46, 47
9744
XM_011537025.2
XP_011535327.1
47, 48
9966
XM_011537026.2
XP_011535328.1
47, 48
10459
XM_011537027.2
XP_011535329.1
47, 48
7484
XM_011537028.2
XP_011535330.1
46, 47
7483
XM_011537029.2
XP_011535331.1
38, 39
6432
XM_017021506.1
XP_016876995.1
47, 48
9864
XM_017021507.1
XP_016876996.1
46, 47
9849
XM_017021508.1
XP_016876997.1
46, 47
9816
XM_017021509.1
XP_016876998.1
44, 45
9807
XM_017021510.1
XP_016876999.1
45, 46
9783
XM_017021511.1
XP_016877000.1
42, 43
6760
XM_017021512.1
XP_016877001.1
46, 47
7451
XM_017021513.1
XP_016877002.1
45, 46
9750
XM_017021514.1
XP_016877003.1
45, 46
9699
XM_017021515.1
XP_016877004.1
47, 48
7365
XM_017021516.1
XP_016877005.1
44, 45
9633
XM_017021517.1
XP_016877006.1
44, 45
9474
XM_017021518.1
XP_016877007.1
41, 42
7888
XM_017021519.1
XP_016877008.1
37, 38
6982
XM_017021520.1
XP_016877009.1
31, 32
5287
GenBank mRNA
AB037830.2
-
43, 44
6687
Back To Top
More Information
Gene Ontology Categories:
Function(s)
ion transmembrane transport
multicellular organism growth
behavioral response to ethanol
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-qlhls:80/10.239.162.255:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0