Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPB41L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184937.1 | NP_001171866.1 | 9 | 839 | ||
NM_001184938.2 | NP_001171867.1 | 9 | 1129 | |||
NM_001184939.1 | NP_001171868.1 | 9 | 839 | |||
NM_020909.3 | NP_065960.2 | 9 | 839 | |||
NR_135920.1 | 9 | 839 | ||||
XM_006712651.1 | XP_006712714.1 | 9 | 819 | |||
XM_006712652.1 | XP_006712715.1 | 9 | 1134 | |||
XM_006712653.1 | XP_006712716.1 | 9 | 839 | |||
XM_011511529.2 | XP_011509831.1 | 9 | 819 | |||
XM_017004567.1 | XP_016860056.1 | 9 | 675 | |||
XM_017004568.1 | XP_016860057.1 | 9 | 839 | |||
GenBank mRNA | AB046768.1 | - | 2 | 88 | ||
AK023019.1 | - | 9 | 790 | |||
AK290895.1 | - | 9 | 749 | |||
BC032822.2 | - | 9 | 1129 | 4827274 | ||
BC054508.1 | - | 9 | 774 | 5502663 |