Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021229.3 | NP_067052.2 | 2 | 922 | ||
GenBank mRNA | AF278532.1 | - | 2 | 924 | ||
AF297711.1 | - | 2 | 774 | |||
AK226070.1 | - | 2 | 779 | |||
AK290591.1 | - | 2 | 459 | |||
AK315796.1 | - | 2 | 781 | |||
AL833767.1 | - | 1 | 326 | |||
AY330211.1 | - | 2 | 473 | |||
BC136798.1 | - | 2 | 897 | 9020788 | ||
BC136799.1 | - | 2 | 897 | 9020789 |