Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165417.1 | NP_001158889.1 | 5, 6 | 869 | ||
NM_001165418.1 | NP_001158890.1 | 4, 5 | 749 | |||
NM_003562.4 | NP_003553.2 | 5, 6 | 902 | |||
GenBank mRNA | AF070548.1 | - | 5, 6 | 756 | ||
AK290376.1 | - | 5, 6 | 653 | |||
AK313010.1 | - | 5, 6 | 758 | |||
BC006508.2 | - | 5, 6 | 758 | 2960933 | ||
BC006519.2 | - | 5, 6 | 758 | 2960933 | ||
BC016294.2 | - | 5, 6 | 677 | 3911858 | ||
BC017170.2 | - | 5, 6 | 758 | 2960933 | ||
BI752999.1 | - | 4, 5 | 524 | |||
BM047391.1 | - | 5, 6 | 617 | |||
CR456834.1 | - | 5, 6 | 645 |