Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012134.2 | NP_036266.2 | 3 | 3363 | ||
GenBank mRNA | AK126474.1 | - | 3 | 2902 | ||
BC080187.1 | - | 3 | 3312 | 6177902 | ||
X54162.1 | - | 3 | 3243 |