Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012180.2 | NP_036312.2 | 4 | 1338 | ||
XM_017008020.1 | XP_016863509.1 | 4 | 602 | |||
GenBank mRNA | AF115514.1 | - | 4 | 722 | ||
AF174596.1 | - | 4 | 554 | |||
AF201932.1 | - | 4 | 747 | |||
AF233224.1 | - | 4 | 593 | |||
AK026319.1 | - | 4 | 761 | |||
AK314483.1 | - | 4 | 756 | |||
AY358721.1 | - | 4 | 724 | |||
BC014679.2 | - | 4 | 740 | 3857760 | ||
BC014903.1 | - | 4 | 1338 | |||
BC040456.1 | - | 5 | 631 | 4281341 | ||
KU178588.1 | - | 3 | 488 | |||
KU178589.1 | - | 4 | 618 |