Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other R3HCC1L SIRNA Assays ›
Gene Symbol:
R3HCC1L
Gene Name:
R3H domain and coiled-coil containing 1 like
Gene Aliases:
C10orf28, GIDRP86, GIDRP88, PSORT
Chromosome Location:
Chr.10: 98134608 - 98244897 on Build GRCh38
UniGene ID:
Hs.419800
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
134375
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: R3HCC1L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256619.1
NP_001243548.1
5
853
NM_001256620.1
NP_001243549.1
4
802
NM_014472.4
NP_055287.4
4
798
NM_138469.2
NP_612478.2
5
895
XM_011539639.1
XP_011537941.1
5
1110
XM_011539640.1
XP_011537942.1
6
915
XM_011539641.1
XP_011537943.1
5
867
XM_011539642.1
XP_011537944.1
6
1207
XM_011539643.1
XP_011537945.1
4
794
XM_011539644.1
XP_011537946.1
4
798
XM_011539645.2
XP_011537947.1
4
1041
XM_011539646.1
XP_011537948.1
3
673
XM_011539647.1
XP_011537949.1
3
675
XM_011539648.1
XP_011537950.1
2
548
XM_011539649.1
XP_011537951.1
5
1110
XM_017016074.1
XP_016871563.1
4
2006
XM_017016075.1
XP_016871564.1
2
548
XM_017016076.1
XP_016871565.1
5
842
XM_017016077.1
XP_016871566.1
7
1005
XM_017016078.1
XP_016871567.1
6
927
XM_017016079.1
XP_016871568.1
6
941
XM_017016080.1
XP_016871569.1
4
847
XM_017016081.1
XP_016871570.1
5
929
XM_017016082.1
XP_016871571.1
3
721
XM_017016083.1
XP_016871572.1
4
737
GenBank mRNA
AF050198.1
-
4
790
AF525304.1
-
5
806
AK292546.1
-
4
797
BC098268.1
-
5
836
40026913
BC098359.1
-
4
739
40026912
BC099848.1
-
4
739
40026916
Back To Top
More Information
Gene Ontology Categories:
Process(es)
nucleotide binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-8dfkb:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0