Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCAF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015397.3 | NP_056212.1 | 9 | 3371 | ||
XM_011517843.1 | XP_011516145.1 | 9 | 3198 | |||
GenBank mRNA | AB067479.1 | - | 9 | 3296 | ||
AF131771.1 | - | 4 | 2237 | |||
AK000354.1 | - | 9 | 3152 | |||
AL117646.1 | - | 6 | 2509 | |||
BC008893.2 | - | 8 | 2939 | |||
BC063823.1 | - | 9 | 3294 | 6184796 | ||
DQ518909.1 | - | 9 | 3296 |