Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CDKL3 SIRNA Assays ›
Gene Symbol:
CDKL3
Gene Name:
cyclin dependent kinase like 3
Gene Aliases:
NKIAMRE
Chromosome Location:
Chr.5: 134298424 - 134371159 on Build GRCh38
UniGene ID:
Hs.719926
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
1344
Purification
HPLC
Standard
In-Vivo Ready
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CDKL3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001113575.1
NP_001107047.1
9
1229
NM_001300853.1
NP_001287782.1
8
995
NM_016508.3
NP_057592.2
9
1229
XM_011543429.2
XP_011541731.1
9
1580
XM_011543439.2
XP_011541741.1
6
733
XM_011543444.2
XP_011541746.1
5
562
XM_011543445.2
XP_011541747.1
5
511
XM_011543447.2
XP_011541749.1
4
422
XM_017009524.1
XP_016865013.1
9
2057
XM_017009525.1
XP_016865014.1
10
1625
XM_017009526.1
XP_016865015.1
10
1623
XM_017009527.1
XP_016865016.1
9
2021
XM_017009528.1
XP_016865017.1
8
2012
XM_017009529.1
XP_016865018.1
9
2057
XM_017009530.1
XP_016865019.1
9
2057
XM_017009531.1
XP_016865020.1
9
2057
XM_017009532.1
XP_016865021.1
8
1975
XM_017009533.1
XP_016865022.1
8
2011
XM_017009534.1
XP_016865023.1
8
2011
XM_017009535.1
XP_016865024.1
8
1975
XM_017009536.1
XP_016865025.1
6
1770
XM_017009537.1
XP_016865026.1
7
1175
XM_017009538.1
XP_016865027.1
9
1065
XM_017009539.1
XP_016865028.1
8
886
XM_017009540.1
XP_016865029.1
7
696
XM_017009541.1
XP_016865030.1
8
1222
XM_017009542.1
XP_016865031.1
7
653
XM_017009543.1
XP_016865032.1
7
680
XM_017009544.1
XP_016865033.1
7
931
XM_017009545.1
XP_016865034.1
6
738
XM_017009546.1
XP_016865035.1
6
767
XM_017009547.1
XP_016865036.1
4
459
XM_017009548.1
XP_016865037.1
7
1862
GenBank mRNA
AF130372.1
-
8
1110
AK294559.1
-
6
617
AK294740.1
-
5
527
AK297586.1
-
4
389
AK299305.1
-
5
477
AK301802.1
-
8
989
AK310955.1
-
5
467
BC041799.1
-
9
1231
5298279
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
cellular protein modification process
protein phosphorylation
regulation of cell cycle
protein kinase activity
cyclin-dependent protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-zvggx:80/10.239.163.62:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0