Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016327.2 | NP_057411.1 | 10 | 1211 | ||
XM_011530222.2 | XP_011528524.1 | 12 | 1515 | |||
XM_011530224.2 | XP_011528526.1 | 10 | 1317 | |||
XM_011530225.2 | XP_011528527.1 | 11 | 1211 | |||
XM_017028825.1 | XP_016884314.1 | 10 | 1302 | |||
XM_017028826.1 | XP_016884315.1 | 11 | 1360 | |||
XM_017028827.1 | XP_016884316.1 | 10 | 1190 | |||
XM_017028828.1 | XP_016884317.1 | 9 | 1147 | |||
GenBank mRNA | AB013885.1 | - | 10 | 1150 | ||
AF163312.1 | - | 10 | 1151 | |||
AK024277.1 | - | 10 | 2479 | |||
AK314727.1 | - | 10 | 1347 | |||
BC033388.1 | - | 10 | 1108 | |||
BC131703.1 | - | 10 | 1164 | 40121343 | ||
BC139843.1 | - | 10 | 1109 | |||
BX537493.1 | - | 4 | 386 | |||
CR456375.1 | - | 10 | 1211 |