Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NGEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114090.1 | NP_001107562.1 | 13 | 2249 | ||
NM_019850.2 | NP_062824.2 | 15 | 2621 | |||
XM_011510923.2 | XP_011509225.1 | 15 | 2587 | |||
XM_011510924.1 | XP_011509226.1 | 11 | 1620 | |||
XM_011510925.1 | XP_011509227.1 | 11 | 1624 | |||
GenBank mRNA | AK297390.1 | - | 13 | 2246 | ||
AK298315.1 | - | 14 | 2131 | |||
BC031573.1 | - | 15 | 2606 | 5186132 |