Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021044.3 | NP_066382.1 | 3 | 1921 | ||
XM_017019380.1 | XP_016874869.1 | 3 | 1852 | |||
XM_017019381.1 | XP_016874870.1 | 3 | 1569 | |||
GenBank mRNA | BC033507.1 | - | 3 | 1921 | 5169286 | |
HQ607677.1 | - | 3 | 1918 | |||
HQ607678.1 | - | 3 | 1925 |