Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZMYND8 SIRNA Assays ›
Gene Symbol:
ZMYND8
Gene Name:
zinc finger MYND-type containing 8
Gene Aliases:
PRKCBP1, PRO2893, RACK7
Chromosome Location:
Chr.20: 47209214 - 47357780 on Build GRCh38
UniGene ID:
Hs.446240
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
135458
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZMYND8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001281771.2
NP_001268700.1
10
1118
NM_001281772.2
NP_001268701.1
12
1317
NM_001281773.2
NP_001268702.1
11
1264
NM_001281774.2
NP_001268703.1
11
1264
NM_001281775.2
NP_001268704.1
10
1193
NM_001281776.2
NP_001268705.1
10
1193
NM_001281777.2
NP_001268706.1
10
1118
NM_001281778.2
NP_001268707.1
10
1118
NM_001281779.2
NP_001268708.1
11
1324
NM_001281780.2
NP_001268709.1
11
1324
NM_001281781.2
NP_001268710.1
10
1118
NM_001281782.2
NP_001268711.1
10
1118
NM_001281783.2
NP_001268712.1
10
1193
NM_001281784.2
NP_001268713.1
10
1118
NM_012408.5
NP_036540.3
10
1193
NM_183047.3
NP_898868.1
10
1193
NM_183048.3
NP_898869.1
10
1118
XM_005260356.4
XP_005260413.1
10
1230
XM_005260358.2
XP_005260415.1
9
972
XM_005260360.4
XP_005260417.1
10
1695
XM_005260362.2
XP_005260419.1
10
970
XM_005260364.2
XP_005260421.1
10
955
XM_005260366.2
XP_005260423.1
10
986
XM_006723760.3
XP_006723823.1
10
1368
XM_006723762.3
XP_006723825.1
10
1069
XM_006723763.1
XP_006723826.1
10
984
XM_006723766.1
XP_006723829.1
10
928
XM_011528749.2
XP_011527051.1
10
1874
XM_011528751.1
XP_011527053.1
10
978
XM_011528752.1
XP_011527054.1
9
960
XM_011528753.1
XP_011527055.1
11
989
XM_011528754.1
XP_011527056.1
10
916
XM_017027760.1
XP_016883249.1
9
975
XM_017027761.1
XP_016883250.1
10
1441
XM_017027762.1
XP_016883251.1
12
2208
XM_017027763.1
XP_016883252.1
10
1121
XM_017027764.1
XP_016883253.1
10
1640
XM_017027765.1
XP_016883254.1
10
970
XM_017027766.1
XP_016883255.1
10
970
XM_017027767.1
XP_016883256.1
10
1785
XM_017027768.1
XP_016883257.1
10
1045
XM_017027769.1
XP_016883258.1
10
955
XM_017027770.1
XP_016883259.1
11
989
XM_017027771.1
XP_016883260.1
10
955
XM_017027772.1
XP_016883261.1
10
928
XM_017027773.1
XP_016883262.1
10
970
XM_017027774.1
XP_016883263.1
9
948
XM_017027775.1
XP_016883264.1
10
916
XM_017027776.1
XP_016883265.1
10
955
XM_017027777.1
XP_016883266.1
9
948
XM_017027778.1
XP_016883267.1
9
960
XM_017027779.1
XP_016883268.1
9
960
XM_017027780.1
XP_016883269.1
9
975
GenBank mRNA
AB032951.1
-
10
1168
AF454056.1
-
10
1010
AK000275.1
-
10
935
AK122966.1
-
9
814
AK225857.1
-
10
1001
AK294511.1
-
12
1127
AK295747.1
-
10
1077
AK299899.1
-
11
1076
AK309228.1
-
10
987
AL137703.1
-
9
930
BC030721.1
-
7
693
BC136608.1
-
10
949
9020598
BC144289.1
-
10
949
9052809
BC146802.1
-
10
959
8860316
BX641005.1
-
11
1209
DQ082998.1
-
9
974
DQ082999.1
-
9
974
DQ368669.1
-
9
974
DQ368670.1
-
9
974
DQ368671.1
-
9
899
DQ368672.1
-
9
899
DQ368673.1
-
9
974
DQ368674.1
-
9
899
DQ368675.1
-
10
1105
DQ368676.1
-
10
1105
DQ368677.1
-
9
899
DQ368678.1
-
9
899
DQ368679.1
-
9
974
DQ368680.1
-
9
899
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
regulation of transcription, DNA-templated
positive regulation of filopodium assembly
positive regulation of dendritic spine development
modulation of excitatory postsynaptic potential
regulation of postsynaptic density protein 95 clustering
positive regulation of dendritic spine maintenance
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription corepressor activity
chromatin binding
transcription cofactor activity
protein binding
zinc ion binding
histone binding
protein N-terminus binding
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0