Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KATNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007044.3 | NP_008975.1 | 10 | 1524 | ||
XM_005266804.2 | XP_005266861.1 | 10 | 1304 | |||
XM_005266805.3 | XP_005266862.1 | 9 | 1284 | |||
XM_017010207.1 | XP_016865696.1 | 11 | 1614 | |||
XM_017010208.1 | XP_016865697.1 | 9 | 1340 | |||
XM_017010209.1 | XP_016865698.1 | 9 | 1182 | |||
XM_017010210.1 | XP_016865699.1 | 10 | 1309 | |||
GenBank mRNA | AF056022.1 | - | 9 | 1220 | ||
AK292090.1 | - | 10 | 1434 |