Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTN2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197233.2 | NP_001184162.1 | 7 | 2479 | ||
NM_007049.4 | NP_008980.1 | 8 | 2591 | |||
NM_078476.3 | NP_510961.1 | 8 | 2820 | |||
GenBank mRNA | AK093886.1 | - | 1 | 2019 | ||
BC016661.1 | - | 8 | 2763 | 4340392 | ||
BX452202.2 | - | 1 | 470 | |||
BX538050.1 | - | 7 | 2134 | |||
U90543.1 | - | 8 | 2532 |