Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHSY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014918.4 | NP_055733.2 | 2 | 1041 | ||
XM_011521364.2 | XP_011519666.1 | 2 | 580 | |||
GenBank mRNA | AB023207.1 | - | 2 | 1059 | ||
AB071402.1 | - | 2 | 579 | |||
AY358529.1 | - | 2 | 1053 | |||
BC046247.1 | - | 2 | 1048 | 5751618 |