Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DIP2A SIRNA Assays ›
Gene Symbol:
DIP2A
Gene Name:
disco interacting protein 2 homolog A
Gene Aliases:
C21orf106, DIP2
Chromosome Location:
Chr.21: 46458942 - 46570900 on Build GRCh38
UniGene ID:
Hs.189585
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
136791
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DIP2A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146115.1
NP_001139587.1
2
333
NM_001146116.1
NP_001139588.1
2
333
NM_015151.3
NP_055966.2
2
333
NM_206889.2
NP_996772.1
2
333
NM_206890.2
NP_996773.1
2
333
NM_206891.2
NP_996774.1
2
333
XM_011529492.2
XP_011527794.1
2
340
XM_011529501.1
XP_011527803.1
2
340
XM_011529502.1
XP_011527804.1
2
340
XM_017028291.1
XP_016883780.1
2
340
XM_017028292.1
XP_016883781.1
2
340
XM_017028293.1
XP_016883782.1
2
340
XM_017028294.1
XP_016883783.1
2
324
XM_017028295.1
XP_016883784.1
2
324
XM_017028296.1
XP_016883785.1
2
324
XM_017028297.1
XP_016883786.1
2
340
XM_017028298.1
XP_016883787.1
2
324
XM_017028299.1
XP_016883788.1
2
340
XM_017028300.1
XP_016883789.1
2
340
XM_017028301.1
XP_016883790.1
2
340
XM_017028302.1
XP_016883791.1
2
340
XM_017028303.1
XP_016883792.1
2
340
XM_017028304.1
XP_016883793.1
2
340
XM_017028305.1
XP_016883794.1
2
340
GenBank mRNA
AB273729.1
-
2
171
AF490768.1
-
2
238
AK056738.1
-
2
225
BC033718.1
-
2
316
4708615
BC038443.1
-
2
332
5217858
DA326960.1
-
2
334
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
multicellular organism development
metabolic process
negative regulation of gene expression
regulation of apoptotic process
catalytic activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-rxbn5:80/10.239.162.255:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0